Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Helix Elite™ Inactivated Adenovirus 2
SDP

Catalog No. 23004117 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-117 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-004-117 Supplier Microbiologics™ Supplier No. HE0026N
 Restricted Item
Add to Cart
Add to Cart

Includes

Five unassayed lyophilized pellets packaged in individual vials

For use as positive control in molecular applications

● Fully intact
● non-viable process controls
● Quick dissolving lyophilized pellets may be reconstituted in buffer or transport medium
● Can be processed using same protocols as patient sample

Certifications

FDA listed and CE Marked

Specifications

Certifications/Compliance FDA approved and CE Marked
Form Lyophilized Pellet
Product Type AdenoVirus
For Use With (Application) In vitro diagnostic
Includes Five assayed lyophilized pellet packaged in individual vials
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.