Learn More
Microbiologics™ BD Max™ KWIK-STIK MRSA Organism QC Set
SDP
Catalog No. 23017214 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
23-017-214 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-214 Supplier Microbiologics™ Supplier No. 8174
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Staphylococcus aureus (ATCC™ 33591™, ATCC™ 700699™, ATCC™ 43300™, ATCC™ 700698™, NCTC 12493, ATCC™ 6538™, ATCC™ 29213™), Staphylococcus aureus subsp. aureus (NCTC 12973, ATCC™ 33862™) and Staphylococcus epidermidis ATCC™ 51625™
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Convenient packaging options
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab
  • For use with BD MAX™ MRSA Assays
  • For added convenience, a peel-off identification label is included for easy documentation
  • Includes one STIK per strain
Specifications
ATCC® Multiple
Product Type QC Microorganism
For Use With (Application) For use with BD MAX™ MRSA Assays
Includes Staphylococcus aureus (ATCC™ 33591™, ATCC™ 700699™, ATCC™ 43300™, ATCC™ 700698™, NCTC 12493, ATCC™ 6538™, ATCC™ 29213™), Staphylococcus aureus subsp. aureus (NCTC 12973, ATCC™ 33862™) and Staphylococcus epidermidis ATCC™ 51625™
Quantity 10/Pk.