Learn More
Microbiologics™ BD Max™ KWIK-STIK Strep B Organism QC Set
SDP
Catalog No. 23017215 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
23-017-215 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-215 Supplier Microbiologics™ Supplier No. 8173
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Streptococcus agalactiae ((B) ATCC™ 12386™, (B, III) ATCC™ 12403™, (B) ATCC™ 27956™, (B) NCTC 9993), Streptococcus dysagalactiae subsp. equisimilis ((C) ATCC™ 12388™, (C) NCTC 8543), Streptococcus pyogenes (A) ATCC™ 19615™, Streptococcus species ((F,2) ATCC™ 12392™, (B, Ib) ATCC™ 12401™) and Streptococcus anginosus (G) NCTC 10713
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Convenient packaging options
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab
  • For use with BD MAX™ GBS Assay
  • For added convenience, a peel-off identification label is included for easy documentation
  • Includes one STIK per strain
Specifications
ATCC® Multiple
Product Type QC set
For Use With (Application) BD MAX™ GBS Assay
Includes Streptococcus agalactiae ((B) ATCC™ 12386™, (B, III) ATCC™ 12403™, (B) ATCC™ 27956™, (B) NCTC 9993), Streptococcus dysagalactiae subsp. equisimilis ((C) ATCC™ 12388™, (C) NCTC 8543), Streptococcus pyogenes (A) ATCC™ 19615™, Streptococcus species ((F,2) ATCC™ 12392™, (B, Ib) ATCC™ 12401™) and Streptococcus anginosus (G) NCTC 10713
Quantity 10/Pk.