Learn More
Microbiologics™ BD Max™ SA KWIK-STIK Microorganism QC Set
SDP
Catalog No. 23017213 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
14/Pk.
Catalog No. Quantity
23-017-213 14/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-017-213 Supplier Microbiologics™ Supplier No. 8178
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Staphylococcus aureus (ATCC™ BAA-2312™, ATCC™ 29213™, BD 1, BD 9, BD 11, BD 16, BD 19, BD 131, BD 797, BD 2800, BD 2937, BD 2952, BD 3097) and Staphylococcus epidermidis ATCC™ 14990™
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Convenient packaging options
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab
  • For use with BD MAX™ StaphSR and BD MAX™ MRSA XT Assays
  • For added convenience, a peel-off identification label is included for easy documentation
  • Includes one STIK per strain
Specifications
Product Type QC Microorganism
For Use With (Application) BD MAX™ StaphSR and BD MAX™ MRSA XT Assays
Includes Staphylococcus aureus (ATCC™ BAA-2312™, ATCC™ 29213™, BD 1, BD 9, BD 11, BD 16, BD 19, BD 131, BD 797, BD 2800, BD 2937, BD 2952, BD 3097) and Staphylococcus epidermidis ATCC™ 14990™
Quantity 14/Pk.