Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-Hydro ShotÖ Enterococcus faecium ATCCÖ 35667Öå
SDP

Catalog No. 16230483 Shop All Microbiologics Products
This item is not returnable. View return policy
Catalog No. 16-230-483 Supplier Microbiologics™ Supplier No. 0968H
This item is not returnable. View return policy

For culture purposes and QC applications

EZ-Hydro Shot™
  • Ready-to-use quantitative QC microorganism product
  • Designed for water quality testing
  • Lyophilized microorganism pellets are packaged in individual glass vials
  • No pre-incubation steps are necessary for its preparation
  • Pellets instantly dissolve, saving laboratories considerable time and labor
  • Delivers low-level concentration of 20-100 CFU
  • Used for variety of water tests such as membrane filtration, enzyme substrate methods and heterotrophic plate count
Designed for Water Testing:
  • Microbiologics validated using Standard Methods (SM) for membrane filtration and enzyme substrate methods
  • Each pellet provides 20-100 CFU
  • Traceable to culture collection
  • Online Certificate of Analysis
Packaging options:
  • Multiple-Strain Kits containing 3 lyophilized pellets of 3 EPA recommended control strains for Coliform/E. coli testing and Enterococci testing (9 pellets total)
  • Single-Strain Kits containing 6 lyophilized pellets of single strain

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

Biosafety Level 1
Form Lyophilized Pellet
Product Type EZ-Hydro Shot
Includes 6 Vials of single enumerated microorganism (each vial containing 1 lyophilized pellet), instructions for Use, and peel off label
Species Enterococcus faecium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.