Learn More
Microbiologics™ EZ-Hydro ShotÖ Escherichia coli ATCCÖ 11775Öå
SDP
Catalog No. 16230484 Shop All Microbiologics Products
Change view
Click to view available options
Format:
Lyophilized Pellet
Catalog No. Format
16-230-484 Lyophilized Pellet
1 options
This item is not returnable. View return policy
Catalog No. 16-230-484 Supplier Microbiologics™ Supplier No. 0465H
This item is not returnable. View return policy
Includes
6 Vials of single enumerated microorganism (each vial containing 1 lyophilized pellet), instructions for Use, and peel off label
For culture purposes and QC applications
EZ-Hydro Shot™
  • Ready-to-use quantitative QC microorganism product
  • Designed for water quality testing
  • Lyophilized microorganism pellets are packaged in individual glass vials
  • No pre-incubation steps are necessary for its preparation
  • Pellets instantly dissolve, saving laboratories considerable time and labor
  • Delivers low-level concentration of 20-100 CFU
  • Used for variety of water tests such as membrane filtration, enzyme substrate methods and heterotrophic plate count
Designed for Water Testing:
  • Microbiologics validated using Standard Methods (SM) for membrane filtration and enzyme substrate methods
  • Each pellet provides 20-100CFU
  • Traceable to culture collection
  • Online Certificate of Analysis
Packaging options:
  • Multiple-Strain Kits containing 3 lyophilized pellets of 3 EPA recommended control strains for Coliform/E. coli testing and Enterococci testing (9 pellets total)
  • Single-Strain Kits containing 6 lyophilized pellets of single strain
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Biosafety Level 1
Form Lyophilized Pellet
Format Lyophilized Pellet
Product Type EZ-Hydro Shot
Includes 6 Vials of single enumerated microorganism (each vial containing 1 lyophilized pellet), instructions for Use, and peel off label
Specimen Type Escherichia coli