Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-Hydro Shot™ Serratia marcescens ATCCÖ 43862Ö
SDP

Catalog No. 16230489 Shop All Microbiologics Products
Change view
Click to view available options
Format:
Lyophilized Pellet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
16-230-489 Lyophilized Pellet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-230-489 Supplier Microbiologics™ Supplier No. 01271UVV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

6 vials of single enumerated microorganism (each vial containing 1 lyophilized pellet), Instructions for Use, and peel off label

For culture purposes and QC applications

EZ-Hydro Shot™
  • Ready-to-use quantitative QC microorganism product
  • Designed for water quality testing
  • Lyophilized microorganism pellets are packaged in individual glass vials
  • No pre-incubation steps are necessary for its preparation
  • Pellets instantly dissolve, saving laboratories considerable time and labor
  • Delivers low-level concentration of 20-100CFU
  • Used for variety of water tests such as membrane filtration, enzyme substrate methods and heterotrophic plate count
Designed for Water Testing:
  • Microbiologics validated using Standard Methods (SM) for membrane filtration and enzyme substrate methods
  • Each pellet provides 20-100CFU
  • Traceable to culture collection
  • Online Certificate of Analysis
Packaging options:
  • Multiple-Strain Kits containing 3 lyophilized pellets of 3 EPA recommended control strains for Coliform/E. coli testing and Enterococci testing (9 pellets total)
  • Single-Strain Kits containing 6 lyophilized pellets of single strain

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

ATCC® ATCC 43862
Biosafety Level 1
Form Lyophilized Pellet
Format Lyophilized Pellet
Product Type EZ-Hydro Shot
Includes 6 vials of single enumerated microorganism (each vial containing 1 lyophilized pellet), Instructions for Use, and peel off label
Species Serratia marcescens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.