Learn More
Microbiologics™ Helix Elite™ Norovirus GI.1Synthetic RNA
SDP
Catalog No. 23004112 Shop All Microbiologics Products
Change view
Click to view available options
Norovirus GI.1
Catalog No.
23-004-112 Norovirus GI.1
1 options
This item is not returnable. View return policy
Catalog No. 23-004-112 Supplier Microbiologics™ Supplier No. HE0013S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
One vial of dried synthetic DNA or RNA (approximately 100 reactions), one vial of molecular standard water for rehydration and Certificate of Analysis
For use as positive control in molecular applications
  • Synthetic DNA/RNA molecular standard
  • ORF1-ORF2 Junction of Norovirus GI.1
  • Dried RNA preserved with Biomatrica RNAstable™
Specifications
Product Type Synthetic RNA