Learn More
Microbiologics™ Bacillus licheniformis ATCC™ 12759™†
SDP
Catalog No. 23016354 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-016-354 -
23-001-865 2 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-016-354 Supplier Microbiologics™ Supplier No. 0812K
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Bacillus licheniformis in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • ATCC™ 12759™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 12759
Product Type Quality Control Organism
Species Bacillus licheniformis