Learn More
Microbiologics™ Citrobacter braakii ATCC™ 43162™†
SDP
Catalog No. 23016682 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
ATCC 43162
Product Type:
Lyfo-Disk QC Control
Catalog No. ATCC® Product Type
23-016-682 ATCC 43162 Lyfo-Disk QC Control
1 options
Catalog No. 23-016-682 Supplier Microbiologics™ Supplier No. 0478L
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Citrobacter braakii strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single microorganism strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43162™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 43162
Product Type Lyfo-Disk QC Control
Species Citrobacter braakii