Learn More
Enterococcus hirae ATCC™ 8043™†, Microbiologics™
SDP
Catalog No. 23002180 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5/Pk.
Catalog No. Quantity
23-002-180 5/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-002-180 Supplier MicroBioLogics Supplier No. 0650X
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Enterococcus hirae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
KWIK-STIK™ Plus
  • Five self-contained KWIK-STIK units with microorganism limited to two passages from reference culture
  • ATCC™ 8043™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® 8043™*
Product Type Quality Control Organism
Quantity 5/Pk.
Species Enterococcus hirae