Learn More
Escherichia coli ATCC™ 23848™†, Microbiologics™
SDP
Catalog No. 23016518 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
23-016-518 6/Pk.
1 options
Catalog No. 23-016-518 Supplier MicroBioLogics Supplier No. 0203L
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Escherichia coli strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Escherichia coli strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 23848™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 23848
Product Type Lyfo-Disk QC Control
Quantity 6/Pk.
Species Escherichia coli