Learn More
Microbiologics™ Mycoplasma pneumoniae ATCC™ 15531™†
SDP
Catalog No. 23014700 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
23-014-700 6/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-014-700 Supplier Microbiologics™ Supplier No. 0503L
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Mycoplasma pneumoniae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Mycoplasma pneumoniaea strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 15531™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 15531
Product Type Lyfo-Disk QC Control
Quantity 6/Pk.
Species Mycoplasma pneumoniae