Learn More
Microbiologics™ Penicillium chrysogenum ATCC™ 10106™†
SDP
Catalog No. 23016507 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
Lyfo-Disk QC Control
Quality Control Organism
ATCC®:
10106
ATCC 10106
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type ATCC®
23-016-507 Lyfo-Disk QC Control ATCC 10106
23-003-875 Quality Control Organism 10106
23-016-071 Quality Control Organism ATCC 10106
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-016-507 Supplier Microbiologics™ Supplier No. 0178L
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Penicillium chrysogenum strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Penicillium chrysogenum strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 10106™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 10106
Product Type Lyfo-Disk QC Control
Species Penicillium chrysogenum