Learn More
Microbiologics™ Sphingomonas paucimobilis ATCC™ 29837™†
SDP
Catalog No. 23016115 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
23-016-115 6/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-016-115 Supplier Microbiologics™ Supplier No. 0274K
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Sphingomonas paucimobilis strain in a lyophilized pellet, a reservoir of hydrating fluid, and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • ATCC™ 29837™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC 29837
Product Type Quality Control Organism
Quantity 6/Pk.
Species Sphingomonas paucimobilis