Learn More
Microbiologics™ Zygosaccharomyces bailii ATCC™ MYA-4549™†
SDP
Catalog No. 23016006 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
ATCC MYA-4549
MYA-4549
Product Type:
Quality Control Organism
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ATCC® Product Type
23-016-006 ATCC MYA-4549 Quality Control Organism
23-004-078 MYA-4549 Quality Control Organism
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-016-006 Supplier Microbiologics™ Supplier No. 01011K
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Zygosaccharomyces bailii strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
  • ATCC™ MYA-4549™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
ATCC® ATCC MYA-4549
Product Type Quality Control Organism
Species Zygosaccharomyces bailii