Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Klebsiella pneumoniae NCTC 13440
SDP

Catalog No. 23019002 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2/Pk.
6/Pk.
Format:
KWIK-STIK
LYFO-DISK
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
23-019-002 2/Pk. KWIK-STIK
23-019-001 6/Pk. LYFO-DISK
23-019-000 6/Pk. KWIK-STIK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-019-002 Supplier Microbiologics™ Supplier No. 01112P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single strain of Klebsiella pneumoniae in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Streptococcus gallolyticus
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • NCTC 13440

Specifications

ATCC® NCTC 13440
Biosafety Level 2
Certifications/Compliance CE, online certificate analysis
Form Lyophilized Pellet
Format KWIK-STIK
Product Type Quality Control Organism
For Use With (Application) Culture purpose and QC application
Packaging Type KWIK STIK
Quantity 2/Pk.
Species Klebsiella pneumoniae
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.