Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ KWIK-STIK™ Cepheid™ Xpert C. difficile QC Set
SDP

Catalog No. 23001910 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-910 2 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-001-910 Supplier Microbiologics™ Supplier No. 8171
Add to Cart
Add to Cart

Includes

Two KWIK-STIK units including C. difficile ATCC 9689 and C. difficile ATCC 700057

Each set contains a positive and negative control

KWIK-STIK™ Plus

  • Designed to meet the needs of laboratories requiring QC mircoorganisms that are a limited number of passages from the reference culture
  • Microorganism preparation is just two pasages from the reference culture, allowing for more subcultures
  • Conveniently packaged in a KWIK-STIK device featuring a lyophilized microorganism pellet, inoculating swab and reservoir of hydrating fluid
  • Ready to use and affordable
  • A peel-off Certificate of Assay is included for quality control documantation
  • Use with Cepheid systems

Specifications

Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Media Type Kwik-stik™
Product Type QC Microorganism
For Use With (Application) For Cepheid™ system
Includes C. difficile ATCC 9689, C. difficile ATCC 700057
Quantity 2 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.