Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Nanosphere Verigene™ KWIK-STIK™ Gram-Positive Blood Culture Test QC Set
SDP

Catalog No. 23001950 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-950 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-001-950 Supplier Microbiologics™ Supplier No. 5225P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Streptococcus pyogenes NCIMB 13285, Streptococcus agalactiae NCTC 8017, Streptococcus anginosus NCTC 10713, Staphylococcus aureus NCTC 12493, Enterococcus faecalis NCTC 13379, Streptococcus pneumoniae NCIMB 13286, Staphylococcus epidermidis NCIMB 8853, Staphylococcus lugdunensis NCTC 7990, Enterococcus faecium NCTC 12204, Listeria monocytogenes NCTC 10890

Microorganism sets for culture purposes and QC applications

Specifications

Format KWIK-STIK
Media Type Kwik-stik™
Product Type Quality Control Organism
Includes Streptococcus pyogenes NCIMB 13285, Streptococcus agalactiae NCTC 8017, Streptococcus anginosus NCTC 10713, Staphylococcus aureus NCTC 12493, Enterococcus faecalis NCTC 13379, Streptococcus pneumoniae NCIMB 13286, Staphylococcus epidermidis NCIMB 8853, Staphylococcus lugdunensis NCTC 7990, Enterococcus faecium NCTC 12204, Listeria monocytogenes NCTC 10890
Quantity 2/Pk.
Species Multiple
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.