Learn More
Microbiologics™ Staphylococcus aureus ATCC™ BAA-2312™
SDP
Catalog No. 23017210 Shop All Microbiologics Products
Change view
Click to view available options
Packaging Type:
KWIK STIK
LYFO DISK
Product Type:
Quality Control Organism
Quantity:
2/Pk.
6/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Product Type Quantity
23-017-210 KWIK STIK Quality Control Organism 2/Pk.
23-017-209 LYFO DISK Quality Control Organism 6/Pk.
23-017-208 KWIK STIK Quality Control Organism 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-017-210 Supplier Microbiologics™ Supplier No. 01122P
Add to Cart
Add to Cart
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features single strain of Staphylococcus aureus in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in pack of 2 or 6
LYFO DISK™
  • Reference stock culture option; lyophilized pellet containing single strain of Staphylococcus aureus
  • Easy to use; re-hydrate pellet and inoculate
  • Available in re-sealable vial containing 6 pellets
  • NCTC 13406
Specifications
ATCC® ATCC BAA-2312
Biosafety Level 2
Product Type Quality Control Organism
Packaging Type KWIK STIK
Quantity 2/Pk.
Species Staphylococcus aureus