Learn More
Microbiologics™ UNEX Buffer
SDP
Catalog No. 10810300 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
10-810-300 100 mL
1 options
This item is not returnable. View return policy
Catalog No. 10-810-300 Supplier Microbiologics™ Supplier No. MR0501
This item is not returnable. View return policy
Isothiocyanate lysis buffer for preserving samples and universal nucleic acid extraction (UNEX) of DNA and RNA from viruses, bacteria and parasites
Specifications
Product Type UNEX Buffer
Quantity 100 mL