Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Vitek™ 2 AST-GP II QC Set
SDP

Catalog No. 23004799 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-799 2 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-799 Supplier Microbiologics™ Supplier No. 5187P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Staphylococcus aureus subsp. aureus™ BAA-976™†, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™†,Staphylococcus aureus ATCC™ BAA-1026™†, Staphylococcus aureus subsp. aureus ATCC™ 29213™†, Enterococcus faecalis ATCC™ 29212™†, Enterococcus faecalis Escheichia coli Streptococous pheumoniae

Microorganism sets for culture purposes and QC applications

  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Three convenient packaging options

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single microorganism strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Specifications

Certifications/Compliance CE, online certificate of analysis
Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Media Type Kwik-stik™
Product Type Microbiology Media
For Use With (Application) Culture purpose and QC application
Includes Enterococcus faecalis ATCC 29212, Staphylococcus aureus subspecies aureus ATCC BAA-976, Staphylococcus aureus subspecies aureus ATCC BAA-977, Staphylococcus aureus ATCC BAA-1026, Staphylococcus aureus subspecies aureus ATCC 29213
Quantity 2 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.