Learn More
Marketlab Microbiology Workstation Organizer
Catalog No. 22505338
Change view
Click to view available options
Dimensions (L x W x H):
14.25 x 10.25 x 4.75 in.
Catalog No. Dimensions (L x W x H)
22-505-338 14.25 x 10.25 x 4.75 in.
1 options
Catalog No. 22-505-338 Supplier Marketlab Supplier No. 60805
Add to Cart
Add to Cart
Designed specifically to support microbiology workstation organization.
  • Eleven different-sized compartments are excellent for accommodating microbiology essentials
  • Room for the most-common microbiology supplies including transfer pipettes, slides, DI water and saline droppers, a timer, and inoculating loops
  • Keeps supplies within easy reach and improves efficiency
  • Grips on the bottom prevent the Organizer from sliding around
  • Transparent for ease of content management
  • Rugged, lightweight polycarbonate
Specifications
Product Type Organizer
Color Clear
Dimensions (L x W x H) 14.25 x 10.25 x 4.75 in.
Material Heavy-duty Plastic