Learn More
Boca Scientific Inc GeneFix Saliva DNA Microbiome Collectors, Pre-filled with Stabilization Buffer, Pack of 50
Catalog No. 501685617
Encompass
GeneFix Saliva Microbiome DNA Collection device provides an efficient, alternative method to traditional collection and storage of microbiome samples. This device stabilizes microbial DNA in tubes pre-filled with a non-toxic stabilization buffer. The collector has a simple, anti-spill screw-on funnel for saliva delivery directly into the buffer. Microbial DNA is stabilized in the buffer for over 1 year at room temperature, unlike traditional methods that require freezing. The robust sample tubes are fully transport tested to 95KPa to allow mailing and storage. Each tube contains a unique 1D bar and human readable code to enable accurate sample tracking.
Catalog No. 50-168-5617 Supplier Boca Scientific Inc Supplier No. MFX01/50
GeneFix Saliva Microbiome DNA Collection device provides an efficient, alternative method to traditional collection and storage of microbiome samples. This device stabilizes microbial DNA in tubes pre-filled with a non-toxic stabilization buffer. The collector has a simple, anti-spill screw-on funnel for saliva delivery directly into the buffer. Microbial DNA is stabilized in the buffer for over 1 year at room temperature, unlike traditional methods that require freezing. The robust sample tubes are fully transport tested to 95KPa to allow mailing and storage. Each tube contains a unique 1D bar and human readable code to enable accurate sample tracking.