Learn More
New England Biolabs, Inc. NEBNext Microbiome DNA Enrichment Kit – 6 reactions
SDP
Catalog No. NC0514634
Encompass
The NEBNext Microbiome DNA Enrichment Kit facilitates enrichment of microbial DNA from samples containing methylated host DNA (including human), by selective binding and removal of the CpG-methylated host DNA. Importantly, microbial diversity remains intact after enrichment.
Catalog No. NC0514634 Supplier New England Biolabs, Inc. Supplier No. E2612S
The NEBNext Microbiome DNA Enrichment Kit facilitates enrichment of microbial DNA from samples containing methylated host DNA (including human), by selective binding and removal of the CpG-methylated host DNA. Importantly, microbial diversity remains intact after enrichment.