Learn More
Cayman Chemical MIcrobIome Metaolt ScrenI 50uL
SDP
Catalog No. 502912935
Encompass_Preferred
A liminoid degradant has antifeedant activity against and inhibits development of armyworm larvae when applied to fresh wheat leaves at 5 10 or 20 mg/ml reduces LPS-induced nuclear translocation of the NF-KB subunit p65 as well as inhibits LPS-induced production of NO and PGE2 in RAW 264 7 cells reduces tumor growth in an A549 mouse xenograft model at 30 or 100 mg/kg
Catalog No. 50-291-2935 Supplier Cayman Chemical Supplier No. 3630550 UL
A liminoid degradant has antifeedant activity against and inhibits development of armyworm larvae when applied to fresh wheat leaves at 5 10 or 20 mg/ml reduces LPS-induced nuclear translocation of the NF-KB subunit p65 as well as inhibits LPS-induced production of NO and PGE2 in RAW 264 7 cells reduces tumor growth in an A549 mouse xenograft model at 30 or 100 mg/kg