Learn More
Thermo Scientific Dionex™ IonPac™ AS30 Analytical and Guard IC Column, Microbore Separator
Catalog No. NC1749239 Shop All Thermo Scientific Products
Encompass
Achieve high resolution separations of trace oxyhalides and inorganic anions in drinking water with Thermo Scientific™ Dionex™ IonPac™ AS30 columns. The column selectivity has been optimized for trace level analysis of bromate in drinking water preserved with ethylenediamine (EDA). EDA is used as a preservative in some drinking water samples and can cause interference with early eluting peaks. The high capacity of this hydroxide-selective anion-exchange column allows large loop injections for trace analysis (µg/L) without sample pre-treatment or preconcentration.
Catalog No. NC1749239 Supplier Thermo Scientific Supplier No. 303161
Achieve high resolution separations of trace oxyhalides and inorganic anions in drinking water with Thermo Scientific™ Dionex™ IonPac™ AS30 columns. The column selectivity has been optimized for trace level analysis of bromate in drinking water preserved with ethylenediamine (EDA). EDA is used as a preservative in some drinking water samples and can cause interference with early eluting peaks. The high capacity of this hydroxide-selective anion-exchange column allows large loop injections for trace analysis (µg/L) without sample pre-treatment or preconcentration.