Learn More
AVANTOR MASTERFLEX (VWR LLC) Microbore tubing, 0.030"" x 0.090""OD, 100 ft/roll
Catalog No. NC1071411
Encompass
Microbore Tubing, Translucent, 0.030" x 0.090"OD, 100 ft/roll, Formulation: Tygon ND-100-80Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.Certifications: Resin meets USP Class VI requirements.Temperature range: ¿25 to 185¿F (¿31 to 85¿C)Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC1071411 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641903
Microbore Tubing, Translucent, 0.030" x 0.090"OD, 100 ft/roll, Formulation: Tygon ND-100-80Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.Certifications: Resin meets USP Class VI requirements.Temperature range: ¿25 to 185¿F (¿31 to 85¿C)Sterilize: Autoclave, ethylene oxide, or gamma irradiation.