Learn More
CELLTREAT Scientific Products 2.0mL Micro Centrifuge Tube, Non-sterile
SDP
Catalog No. NC1964639
Encompass_Preferred
All tubes have a frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand.
Catalog No. NC1964639 Supplier CELLTREAT Scientific Products Supplier No. 230403
All tubes have a frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand.