Learn More
RWD LIFE SCIENCE INC MICROCENTRIFUGE
SDP
Catalog No. NC3404385
Encompass_Preferred
M1324 microcentrifuge is a ventilated model, satisfying most of the common standard applications, Even at the highest speed, it can still work stably and quietly. Also, the compact-design microcentrifuge not only equips a standard 24-place quick-lock rotor (closed with only 1/6 turn) but also adapt PCR tubes and spin column, satisfying different customers’ needs.The microcentrifuge enables your centrifugation simple by providing 10 acceleration and deceleration ramps . Featured with 48 user-defined programs provide quick and convenient setting and parameter use.
Catalog No. NC3404385 Supplier RWD LIFE SCIENCE INC Supplier No. M1324
M1324 microcentrifuge is a ventilated model, satisfying most of the common standard applications, Even at the highest speed, it can still work stably and quietly. Also, the compact-design microcentrifuge not only equips a standard 24-place quick-lock rotor (closed with only 1/6 turn) but also adapt PCR tubes and spin column, satisfying different customers’ needs.The microcentrifuge enables your centrifugation simple by providing 10 acceleration and deceleration ramps . Featured with 48 user-defined programs provide quick and convenient setting and parameter use.