Learn More
Chemglass Life Sciences Micro Centrifuge Cool Block, 90 Well
SDP
Catalog No. 501532368
Encompass
Micro Centrifuge Cool Block, 90 Well. Large 90 well metal chamber for 1.5-2 mL centrifuge tubes. This chamber is made of solid aluminum and the wells are made for snug fit for the tubes. Place on ice to chill large number of tubes. The metal chamber provides a very stable platform in an ice bucket or can be put on hot plate to heat large number of sample tubes.
Catalog No. 50-153-2368 Supplier Chemglass Life Sciences Supplier No. CLS431201
Micro Centrifuge Cool Block, 90 Well. Large 90 well metal chamber for 1.5-2 mL centrifuge tubes. This chamber is made of solid aluminum and the wells are made for snug fit for the tubes. Place on ice to chill large number of tubes. The metal chamber provides a very stable platform in an ice bucket or can be put on hot plate to heat large number of sample tubes.