Learn More
Sigma Aldrich Fine Chemicals Biosciences Microcentrifuge Tube natur
Catalog No. 502900941
Encompass
Microcentrifuge Tubes are certified DNase RNase and Pyrogen free.
Catalog No. 50-290-0941 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS4323500EA
Microcentrifuge Tubes are certified DNase RNase and Pyrogen free.