Learn More
Bellco Glass, Inc. Microcentrifuge Tube Rack 96 P
SDP
Catalog No. 502330792
Encompass_Preferred
Microcentrifuge Tube Racks are reversible 96-well racks that hold tubes in an 8 × 12 format. One side of the reversible rack holds 96 1.5 mL, 1.8 mL or 2.0 mL microcentrifuge tubes while the other side holds 96 0.5 mL tubes.Racks are alpha-numerically labeledDesigned to be stackable and are autoclavableLids are sold separatelyDimensions: 9" (L) × 4½" (W) × 1" (H) excluding lid. Color-Blue, Material-Polypropylene (PP), Package Type-Bulk, Carton Qty-5
Catalog No. 50-233-0792 Supplier Bellco Glass, Inc. Supplier No. 130000372
Microcentrifuge Tube Racks are reversible 96-well racks that hold tubes in an 8 × 12 format. One side of the reversible rack holds 96 1.5 mL, 1.8 mL or 2.0 mL microcentrifuge tubes while the other side holds 96 0.5 mL tubes.Racks are alpha-numerically labeledDesigned to be stackable and are autoclavableLids are sold separatelyDimensions: 9" (L) × 4½" (W) × 1" (H) excluding lid. Color-Blue, Material-Polypropylene (PP), Package Type-Bulk, Carton Qty-5