Learn More
Revvity Health Sciences Inc Microcentrifuge Tube Rotor with Aerosol Tight Lid, 16 x 5 mL
Catalog No. 502809944
Encompass
Microcentrifuge Tube Rotor with Aerosol Tight Lid, 16 x 5 mL
Catalog No. 50-280-9944 Supplier Revvity Health Sciences Inc Supplier No. 403055
Microcentrifuge Tube Rotor with Aerosol Tight Lid, 16 x 5 mL