Learn More
Andwin Scientific Micro Centrifuge Tube with Flat Screw-Cap, 1.5 mL, Sterile, 500/pk
SDP
Catalog No. NC1133932
Encompass_Preferred
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents. They feature universal screw threads for use with most popular brand screw caps. Low-profile caps include an ethylene propylene O-ring, ensuring a secure seal.
Catalog No. NC1133932 Supplier Andwin Scientific Supplier No. 16466046
These tubes are made from ultra-clear USP Class VI PP that allows easy viewing of contents. They feature universal screw threads for use with most popular brand screw caps. Low-profile caps include an ethylene propylene O-ring, ensuring a secure seal.