Learn More
Pipette Com BenchMate Microcentrifuge Tubes - 1.7mL (1.5mL compatible) Graduated, Sterile, Low-Retention, in Bulk Packaging. 500/pk
SDP
Catalog No. NC3356532
Encompass
These BenchMate Microcentrifuge Tubes by Oxford Lab Products are essential for your laboratory needs. The 1.7mL capacity (compatible with 1.5mL), graduated tubes come in a bulk packaging of 500 tubes per pack. The tubes are sterile, ensuring reliable and contamination-free use in various applications. Additionally, the low-retention feature minimizes sample wastage, making these tubes highly efficient for your experiments. The convenient graduation marks on the tubes allow for easy and accurate volume measurements. These high-quality microcentrifuge tubes are a valuable addition to any lab, offering precision and consistency for your daily work. Whether you are working with samples, reagents, or enzymes, these tubes provide a dependable solution for your centrifugation needs. Obtain optimal results and streamline your processes with these BenchMate Microcentrifuge Tubes.
Catalog No. NC3356532 Supplier Pipette Com Supplier No. OMT017BSL
These BenchMate Microcentrifuge Tubes by Oxford Lab Products are essential for your laboratory needs. The 1.7mL capacity (compatible with 1.5mL), graduated tubes come in a bulk packaging of 500 tubes per pack. The tubes are sterile, ensuring reliable and contamination-free use in various applications. Additionally, the low-retention feature minimizes sample wastage, making these tubes highly efficient for your experiments. The convenient graduation marks on the tubes allow for easy and accurate volume measurements. These high-quality microcentrifuge tubes are a valuable addition to any lab, offering precision and consistency for your daily work. Whether you are working with samples, reagents, or enzymes, these tubes provide a dependable solution for your centrifugation needs. Obtain optimal results and streamline your processes with these BenchMate Microcentrifuge Tubes.