Learn More
Crystalgen Inc Microcentrifuge Tubes 2.0 mL
SDP
Catalog No. NC1811375
Encompass_Preferred
Crystalgen microcentrifuge tubes are easy to use, and designed to prevent evaporation, leakage, and accidental opening. Audible open & close pops seal tubes for boiling, freezing, centrifugation, and exposure to phenol/chloroform mixtures. The tube thickness is optimized for optical clarity and strength. Tubes are manufactured from 100% virgin polypropylene. Certified free of detectable RNase, DNase, DNA and pyrogens.
Catalog No. NC1811375 Supplier Crystalgen Inc Supplier No. L1192
Crystalgen microcentrifuge tubes are easy to use, and designed to prevent evaporation, leakage, and accidental opening. Audible open & close pops seal tubes for boiling, freezing, centrifugation, and exposure to phenol/chloroform mixtures. The tube thickness is optimized for optical clarity and strength. Tubes are manufactured from 100% virgin polypropylene. Certified free of detectable RNase, DNase, DNA and pyrogens.