Learn More
Biogenex Laboratories Microchamber Slides, 1 case (20 boxes) of OptiPlus™ Positive-Charged Micro Chamber Slides (2/3 test area)
SDP
Catalog No. NC3727592
Encompass_Preferred
OptiPlus™ slides have a positive charge, which allows strong adhesion for tissues and cells. All slides feature a frosted write-on area.
Catalog No. NC3727592 Supplier Biogenex Laboratories Supplier No. XT013CL
OptiPlus™ slides have a positive charge, which allows strong adhesion for tissues and cells. All slides feature a frosted write-on area.