Learn More
KAYAKU ADVANCED MATERIALS INC MICROCHEM Remover PG 16L (4 x 4L) case.
Catalog No. NC1258145
Encompass
MicroChem Remover: Clean Efficient Removal - Residue-free removal with less agitation. AQUEOUS ALKALINE, METAL ION FREE. Reduced device contamination. Rinses completely in water. No strong objectionable odor; Simple replenishment; Cost effective two bath system
Catalog No. NC1258145 Supplier KAYAKU ADVANCED MATERIALS INC Supplier No. G050200 16L4PECS
MicroChem Remover: Clean Efficient Removal - Residue-free removal with less agitation. AQUEOUS ALKALINE, METAL ION FREE. Reduced device contamination. Rinses completely in water. No strong objectionable odor; Simple replenishment; Cost effective two bath system