Learn More
Beckman Coulter MicroClime Environmental Lid, 10/pk
Catalog No. NC1732006 Shop All Beckman Coulter Products
Encompass
The MicroClime Environmental Lid minimizes edge effects and preserves the concentrations of solutions in microplate wells.
Catalog No. NC1732006 Supplier Beckman Coulter Supplier No. 0015719
The MicroClime Environmental Lid minimizes edge effects and preserves the concentrations of solutions in microplate wells.