Learn More
Beckman Coulter Environmental Lid, 50/cs
Catalog No. NC1732010 Shop All Beckman Coulter Products
Encompass
The MicroClime Environmental Lid minimizes edge effects and preserves the concentrations of solutions in microplate wells.
Catalog No. NC1732010 Supplier Beckman Coulter Supplier No. 0015716
The MicroClime Environmental Lid minimizes edge effects and preserves the concentrations of solutions in microplate wells.