Learn More
MSC BW Technologies by Honeywell GasAlertMicroClip XL, Multi-Gas Detector, 1 ea
Catalog No. 502886321
Encompass
The GasAlertMicroClip XL is a robust personal safety device designed to continuously monitor hazardous gas levels, providing critical warnings when concentrations exceed user-defined alarm setpoints.
  • Detects hydrogen sulfide, carbon monoxide, oxygen, and combustible gases (LEL)
  • Features visual, vibrating, and 95 dB audible alarms for comprehensive notification
  • Offers 18 hours of battery life for extended operational periods
  • Operates effectively in temperatures ranging from -20 to 50°C
  • Rated IP68 for excellent ingress protection against dust and water
  • Includes IntelliFlash for continuous visual confirmation of proper operation
Catalog No. 50-288-6321 Supplier MSC Supplier No. 40281941
The GasAlertMicroClip XL is a robust personal safety device designed to continuously monitor hazardous gas levels, providing critical warnings when concentrations exceed user-defined alarm setpoints.
  • Detects hydrogen sulfide, carbon monoxide, oxygen, and combustible gases (LEL)
  • Features visual, vibrating, and 95 dB audible alarms for comprehensive notification
  • Offers 18 hours of battery life for extended operational periods
  • Operates effectively in temperatures ranging from -20 to 50°C
  • Rated IP68 for excellent ingress protection against dust and water
  • Includes IntelliFlash for continuous visual confirmation of proper operation