Learn More
Zymo Research Corporation Micrococcal Nuclease (10 U /100 μl)
SDP
Catalog No. 501957640
Encompass
Complete digestion with Micrococcal Nuclease yields mono- and oligonucleotides with 3-phosphates.
Catalog No. 50-195-7640 Supplier Zymo Research Corporation Supplier No. D52201
Complete digestion with Micrococcal Nuclease yields mono- and oligonucleotides with 3-phosphates.