Learn More
Takara Bio MICROCOCCAL NUCLEASE 15000U
SDP
Catalog No. 50443733
Encompass
Enzyme; Micrococcal nuclease; 15,000U; Digests single-stranded DNA; Will also digest double-stranded DNA or RNA; Yield 3'-phosphate mononucleotides and oglionucleotides; Requires calcium; Completely inactivated by EDTA and EGTA
Catalog No. 50-443-733 Supplier Takara Bio Supplier No. 2910A
Enzyme; Micrococcal nuclease; 15,000U; Digests single-stranded DNA; Will also digest double-stranded DNA or RNA; Yield 3'-phosphate mononucleotides and oglionucleotides; Requires calcium; Completely inactivated by EDTA and EGTA