Learn More
Medchemexpress LLC Micrococcal nuclease | 9013-53-0 | 5 KU
SDP
Catalog No. 5000400386
Encompass_Preferred
Micrococcal nuclease is an endonuclease derived from Staphylococcus aureus. It can digest both single and double-stranded DNA (ssDNA and dsDNA) and RNA, specifically cleaving and hydrolyzing AT or AU-rich regions. It can also be used as an indicator of Staphylococcus aureus contamination.
  • Solid appearance
  • Soluble in H₂O at a concentration of ≥ 20 mg/mL (saturation unknown)
  • Research use only
Catalog No. 50-004-00386 Supplier Medchemexpress LLC Supplier No. HYP27545KU
Micrococcal nuclease is an endonuclease derived from Staphylococcus aureus. It can digest both single and double-stranded DNA (ssDNA and dsDNA) and RNA, specifically cleaving and hydrolyzing AT or AU-rich regions. It can also be used as an indicator of Staphylococcus aureus contamination.
  • Solid appearance
  • Soluble in H₂O at a concentration of ≥ 20 mg/mL (saturation unknown)
  • Research use only