Learn More
Cayman Chemical MIcrococcIn P1 2.5mg
SDP
Catalog No. 502967972
Encompass_Preferred
A macrocyclic peptide antibiotic that prevents the growth of Gram-positive bacteria without affecting that of Gram-negative bacteria
Catalog No. 50-296-7972 Supplier Cayman Chemical Supplier No. 170932.5 MG
A macrocyclic peptide antibiotic that prevents the growth of Gram-positive bacteria without affecting that of Gram-negative bacteria