Learn More
ATCC Micrococcus luteus (Schroeter) Cohn
Supplier:  ATCC 4698/DEL
Encompass
Type strain. Quality control strain for BBL products. Production of 6-aminopenicillanic acid, and L-aspartyl-L-phenylalanine esters. Used to detect for aerosols, and is sensitive to lysozyme. Genome sequenced strain.
Catalog No. 50-189-161FP