Learn More
ATCC Micrococcus luteus
Catalog No. 502383598
Encompass
This strain of Micrococcus luteus is a whole-genome sequenced type strain with applications in quality control and the detection of aerosols, and it produces 6-aminopenicillanic acid and L-aspartyl-L-phenylalanine esters.
Catalog No. 50-238-3598 Supplier ATCC Supplier No. 4698
This strain of Micrococcus luteus is a whole-genome sequenced type strain with applications in quality control and the detection of aerosols, and it produces 6-aminopenicillanic acid and L-aspartyl-L-phenylalanine esters.