Learn More
ATCC Micrococcus luteus
Supplier:  ATCC 10240
Encompass
Micrococcus luteus strain 130.21 is a whole-genome sequenced bacterium that was isolated from air. This aerobe has applications in quality control, bacitracin assays, antibiotic research, and pharmaceutical and personal care research.
Catalog No. 50-238-4553