Learn More
Epoch Life Science Inc EconoSpin® Micro DNA/RNA Columns, 5ul Elution Volume w/ lid (50 columns with collection tubes)
SDP
Catalog No. 501538119
Encompass_Preferred
Silica membrane micro spin columns with lid and collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 40 ug plasmid DNA. Minimum elution volume: 5 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.
Catalog No. 50-153-8119 Supplier Epoch Life Science Inc Supplier No. 3010050
Silica membrane micro spin columns with lid and collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 40 ug plasmid DNA. Minimum elution volume: 5 ul. Compatible with third party buffer sets. 18 month shelf life. Used by labs around the world.